3-Methylthiophene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

M84402-100G M84402-25G
SIALM84402-100GEA 110 EUR
SIALM84402-100G SIALM84402-25G
3-Methylthiophene, Sigma-Aldrich®
3-Methylthiophene
Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: ∼114 °C (1013 hPa)
Melting Pt: –69 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005470
CAS Number: 616-44-4
EINECS: 210-482-6
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR