4-Methyl-3-thiosemicarbazide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
130060-10G
130060-50G
SIAL130060-10GEA
62.1
EUR
SIAL130060-10G
SIAL130060-50G
4-Methyl-3-thiosemicarbazide, Sigma-Aldrich®
4-Methyl-3-thiosemicarbazide
Formula:
C₂H₇N₃S MW: 105.16 g/mol Boiling Pt: 201 °C (1013 hPa) Melting Pt: 136 °C Density: 1.362 g/cm³ (20 °C) |
MDL Number:
MFCD00007617 CAS Number: 6610-29-3 EINECS: 229-563-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...