4-Phenyl-3-thiosemicarbazide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Phenyl-3-thiosemicarbazide, N-Phenylhydrazinecarbothioamide, 4-Phenylthiosemicarbazide
M-Clarity™ quality level = MQ200
131482-10G
131482-50G
SIAL131482-10GEA
63.3
EUR
SIAL131482-10G
SIAL131482-50G
4-Phenyl-3-thiosemicarbazide, Sigma-Aldrich®
4-Phenyl-3-thiosemicarbazide
Formula:
C₇H₉N₃S MW: 167.23 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 138…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007615 CAS Number: 5351-69-9 EINECS: 226-329-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...