1-Adamantanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol
M-Clarity™ quality level = MQ200
130346-25G 130346-5G
SIAL130346-25GEA 68.6 EUR
SIAL130346-25G SIAL130346-5G
1-Adamantanol, Sigma-Aldrich®
1-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 85…87 °C
Density: 1.06 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074729
CAS Number: 768-95-6
EINECS: 212-202-8