1,3,5-Trioxane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Trioxymethylene, sym. Trioxane, s-Trioxane, Formaldehyde trimer
M-Clarity™ quality level = MQ200
T81108-100G T81108-25G T81108-2KG T81108-500G
SIALT81108-100GEA 46.5 EUR
SIALT81108-100G SIALT81108-25G SIALT81108-2KG SIALT81108-500G
1,3,5-Trioxane, Sigma-Aldrich®
1,3,5-Trioxane
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 114.5 °C (1013 hPa)
Melting Pt: 60…62 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00006563
CAS Number: 110-88-3
EINECS: 203-812-5
UN: 1325
ADR: 4.1,III
Merck Index: 12,09863