Ethyl methyl carbonate 98% (by GC)
Supplier: Merck
Warning
Synonyms: Carbonic acid ethyl methyl ester
752002-100ML 752002-10ML 752002-50ML
SIAL752002-100MLEA 327 EUR
SIAL752002-100ML SIAL752002-10ML SIAL752002-50ML
Ethyl methyl carbonate 98% (by GC)
Ethyl methyl carbonate
Formula: C₄H₈O₃
MW: 104.1045 g/mol
Density: 1.006 g/ml (25 °C)
Flash Pt: 23 °C
MDL Number: MFCD00191398
CAS Number: 623-53-0
UN: 3272
ADR: 3 III
Order Now
Specification Test Results
Infrared Spectrum Conforms to Structure
Purity (GC) ≥97.5%