Autoclavable Bags
Supplier: NeoLab
129-9172EA 60.5 EUR
129-9172 NEOL1-7076 NEOL1-7074 NEOL1-7075 NEOL1-7072
Autoclavable Bags
Bags Waste Disposal Bags
Bags, PP, thickness: 40 - 50 µm
  • Suitable for infectious materials
  • Autoclavable up to 121 °C
  • Transparent