Jolting volumeter
Supplier: Gerber
GEAG09.0220EA 8640 EUR
GEAG09.0220
Jolting volumeter
Density Meters
Volumeter designed for determination of specific weight of a jolted sample.
Automatic apparatus with up to 100 strokes in 30 seconds.