D(+)-Galactosamine hydrochloride ≥98%, crystalline solid
Supplier: Cayman Chemical
Synonyms: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
22981-1 22981-10 22981-25 22981-5
CAYM22981-1EA 59 EUR
CAYM22981-1 CAYM22981-10 CAYM22981-25 CAYM22981-5
D(+)-Galactosamine hydrochloride ≥98%, crystalline solid
D(+)-Galactosamine hydrochloride
D-Galactosamine is an amino sugar derivative of D-galactose. D-Galactosamine is hepatotoxic and is used, alone or in combination with LPS, as a model of liver failure in rodents.
Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135830
CAS Number: 1772-03-8
EINECS: 217-198-1
Order Now
Specification Test Results
Purity ≥98%