Protective hand creams, Physioderm®, SANSIBAL®
Supplier: Faweco Hautschutz
Physioderm®
FAWE13632-003EA 120 EUR
FAWE13632-003
Protective hand creams, Physioderm®, SANSIBAL®
Skincare Skin Creams Hand Creams
The water-soluble, pigment-based emulsion develops a separating and a protective film. This solid grid effectively prevents water-insoluble hazards from penetrating the upper layers of the skin. The protective film also reduces the adherence of dirt.
  • O/W emulsion
  • Slightly greasy cream
  • Silicone-free, perfumed
  • skin-neutral pH value
  • Fast absorbing
Suitable when handling water-insoluble substances, e.g. oil, grease or non-water-based cooling lubricants.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904