DMSO resistant peel heat seal
Supplier: Azenta Life Sciences
FTIT4TI-0585EA 1180 EUR
FTIT4TI-0585 FTIT4TI-0586S FTIT4TI-0586-DMSO FTIT4TI-0587
DMSO resistant peel heat seal
Seals Microplate Seals
Solvent resistant heat sealing foil suitable for low and room temperature compound storage.
  • Peelable from PP and COC plates, forms a weld seal to PE plates
  • Temperature range –80 to +40 °C