Anti-CALB2 Mouse Monoclonal Antibody [clone: 3G9]

Supplier: Biosensis
BSENM-1799-100EA 302 EUR
BSENM-1799-100
Anti-CALB2 Mouse Monoclonal Antibody [clone: 3G9]
Antibodies
Calretinin is a calcium-binding protein which is abundant in auditory neurons. Ref: uniprot.org. Antibody is specific for calbindin and does not recognize closely related proteins parvalbumin, calretinin and secretagogin as determined by Western Blotting.

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB2
Clonality: Monoclonal
Clone: 3G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR