ESD safety shoes, lace-up, Biomex Dynamics, Impulse Green
Supplier: ELTEN
722551/36 722551/37 722551/38 722551/39 722551/40 722551/41 722551/42 722551/43 722551/44 722551/45 722551/46
ELTN722551/36EA 86.9 EUR
ELTN722551/36 ELTN722551/37 ELTN722551/38 ELTN722551/39 ELTN722551/40 ELTN722551/41 ELTN722551/42 ELTN722551/43 ELTN722551/44 ELTN722551/45 ELTN722551/46
ESD safety shoes, lace-up, Biomex Dynamics, Impulse Green
Shoes and Boots
Metal- and leather-free, lightweight ESD safety shoe made from perforated microfibre and reflective materials, skin friendly textile lining, a full padded bellows tongue and collar plus an extremely flexible, egronomic S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
  • Climate-regulating, breathable textile lining
  • Anatomically-shaped insole supports the natural heel-to-toe foot movement
  • Replaceable insole; orthopaedic insoles possible
  • Full length ESD inlay sole
  • Abrasion-proof scuff cap
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/green
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.