ESD safety shoes, lace-up, Safety-Grip, White Loop
Supplier: ELTEN
72016/39 72016/40 72016/41 72016/42 72016/43 72016/44 72016/45 72016/46 72016/47 72016/48 72016/49 72016/50
114-8525EA 72.9 EUR
114-8525 114-8526 114-8527 114-8528 114-8529 114-8530 114-8531 114-8532 114-8533 114-8534 114-8535 114-8536
ESD safety shoes, lace-up, Safety-Grip, White Loop
Shoes and Boots
Leather-free ESD safety shoe made from hydrophobised microfibre, a padded tongue and a MONO-PU sole SAFETY-GRIP.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Soft fleece ESD insole
  • Anatomically shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: White
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC