N-Acetyl-L-isoleucine 98% (by HPLC)
Supplier: Thermo Scientific
Synonyms: Ac-Ile-OH, N-Acetyl-L-Isoleucine, (2S,3S)-2-Acetamido-3-methylpentanoic acid
H66771.06 H66771.14
H66771.06EA 80 EUR
H66771.06 H66771.14
N-Acetyl-L-isoleucine 98% (by HPLC)
N-Acetyl-L-isoleucine
Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066072
CAS Number: 3077-46-1
EINECS: 221-364-9
Order Now
Specification Test Results
Assay (HPLC) 97.5%