Titanium, gauze_wire Ø 0.05 mm (0.002 in) 100 mesh
Supplier: Thermo Fisher Scientific
46406.FL 46406.GT
46406.FLEA 154 EUR
46406.FL 46406.GT
Titanium, gauze_wire Ø 0.05 mm (0.002 in) 100 mesh
Titanium
Wire Cloth
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610