Bloom test vessel
Supplier: DWK Life Sciences
646-0035EA 75.83 EUR
646-0035
Bloom test vessel
Reaction Vessels Reaction Vessels, Beaker Shape
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Manufactured according to DIN ISO 9665.