Vacuum oven, WOV
Supplier: Witeg
WITGDH.WOV03020EA 3833.77 EUR
WITGDH.WOV03020 WITGDH.WOV03070
Vacuum oven, WOV
Ovens Vacuum Ovens
Ideal for drying, baking, conditioning, curing, out-glassing solids and liquids, vacuum embedding, moisture testing, plating and aging test.
  • Easy and convenient for vacuuming and venting
  • Easy to read vacuum gauge
  • Superior uniformity and stability
  • Stainless steel interior, powder-coated steel body and glass wool insulation
  • High quality door sealing
  • Storage function for set values of alarm and timer
  • High quality illuminated LCD
Safety mechansims like locking mode, overheat and over current protection, sensor error detection and leakage breaker.