Phenyl-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier: Thermo Fisher Scientific
Synonyms:
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-phenoxytetrahydro-2H-pyran-3,4,5-triol, Phenyl β-D-Glucoside
J66885.14
J66885.06
J66885.14EA
301
EUR
J66885.14
J66885.06
Phenyl-β-D-glucopyranoside ≥98.0% (by HPLC)
Phenyl β-D-glucopyranoside
A substrate used for the determination of D-glucosidase.
Formula:
C₁₂H₁₆O₆ Melting Pt: 173...176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064089 CAS Number: 1464-44-4 EINECS: 215-978-6 |
Specification Test Results
Assay (HPLC) | Min. 98.0% |
Optical Rotation (C=1 in Water 20 °C) | –73 ±2 °C |
Melting Point | 170...176 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...