Nitric acid 10%, StandARd®, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
Danger
H262-43EA 135.6 EUR
H262-43 H262-01
Nitric acid 10%, StandARd®, Macron Fine Chemicals™
Nitric acid
Formula: HNO₃
MW: 63,01 g/mol
Density: 1,05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011349
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (HNO₃)(g/100 mL) 9.5 - 10.5