Capillary columns, Rtx®-5
Supplier: Restek
Rtx®
REST10223-6850EA 740 EUR
REST10223-6850 REST10287-304 REST10284-6850 REST10239-365 REST10257-362 REST10239-324 REST10253-6850 REST10226-303 REST10239-325 REST10252-305 REST10226-304 REST10238-271 REST10253-303 REST10285-6850 REST40201-6850 REST10270-304 REST10270-305 REST10279-305 REST10254-6850 REST10238-6850 REST10254-280 REST10224-125042 REST10224-042 REST10283-6850 REST10224-6850 REST10254-328 REST10254-329 REST10267-6850 REST10255-042 REST10277-305 REST10251-305 REST10279-6850 REST10282-305 REST10279-1266850 REST10290-6850 REST10256-6850 REST10224-128042 REST10239-304 REST10239-303 REST10283-305 REST10223-303 REST10239-305 REST10251-6850 REST10285-1266850 REST10257-6850 REST10220-6850 REST10284-042 REST10252-6850 REST10254-303 REST10284-304 REST10254-304 REST10224-304 REST10254-305 REST10254-307 REST10285-305 REST10272-304 REST10239-6850 REST10255-305 REST10254-031 REST10238-303 REST10224-1286850
Capillary columns, Rtx®-5
Chromatography Columns
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles.
Low polarity phase; Crossbond® 5% diphenyl/95% dimethyl polysiloxane
Temperature range: –60 to 350 °C
Suitable replacement for: HP-5, DB-5, CP-Sil 8 CB, ZB-5 NOTE: DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS. Equivalent to USP G27 and G36 phases.