Diesel fuel #2 composite standard
Supplier: Restek
31258 31259
REST31258EA 79.5 EUR
REST31258 REST31259
Diesel fuel #2 composite standard
Standards Petroleum Industry Standards
Diesel fuel composite standards are comprised of three separate sources blended in equal portions. It may exhibit lot-to-lot variation.
Certifikace: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements