FAME #13 mix
Supplier: Restek
REST35034EA 151 EUR
REST35034
FAME #13 mix
Standards Food Standards
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for mustard seed oil
This mix contains fourteen components such as C16:00 methyl palmitate (3%), C16:01 methyl palmitoleate (1%), C18:00 methyl stearate (2%), C18:01 methyl oleate (20%), C18:02 methyl linoleate (15%), C18:03 methyl linolenate (10%), C20:00 methyl arachidate (1%), C20:01 methyl eicosenoate (10%), C20:02 methyl eicosadienoate (2%), C22:00 methyl behenate (1%), C22:01 methyl erucate (30%), C22:02 methyl docosadienoate (2%), C24:00:00 methyl lignocerate (1%), C24:01:00 methyl nervonate, (2%).