FAME #15 mix
Supplier: Restek
REST35036EA 138 EUR
REST35036
FAME #15 mix
Standards Food Standards
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for peanut oil
This mix contains eight components such as C16:00 methyl palmitate (10%), C18:00 methyl stearate (3%) C18:01 methyl oleate (50%), C18:02 methyl linoleate (30%), C20:00 methyl arachidate (1,5%), C20:01 methyl eicosenoate (1,5%), C22:00 methyl behenate (3%), C24:00:00 methyl lignocerate (1%).