Ethylenedioxybis(trimethylsilane) 98% (by GC)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane
Packaged under nitrogen.
H63857.09 H63857.18
H63857.09EA 47.2 EUR
H63857.09 H63857.18
Ethylenedioxybis(trimethylsilane) 98% (by GC)
Ethylenedioxybis(trimethylsilane)
Formula: C₈H₂₂O₂Si₂
MW: 206.43 g/mol
Boiling Pt: 165…171 °C
Density: 0.842 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009640
CAS Number: 7381-30-8
EINECS: 230-950-3
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Assay (GC) >97.5%