Zinc carbonate, white powder
Supplier: MP Biomedicals
0210473780 0210473790
ICNA0210473780EA 54.6 EUR
ICNA0210473780 ICNA0210473790
Zinc carbonate, white powder
Zinc carbonate
Zinc arbonate is a mineral ore of zinc.
  • Refractive Index: 1.82 (lit.)
Upozornení: Research use only (RUO)
Formula: CO₃Zn
Density: 4.398 g/cm³ (68 °F) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00041957
CAS Number: 3486-35-9
EINECS: 222-477-6
Order Now
Specification Test Results
Identity Test Passes