Arsenic limit test apparatus
Supplier: NEUBERT VOLUME GLASSWAERE
2390-100-19 2390-100-29
NEUB2390-100-19EA 363 EUR
NEUB2390-100-19 NEUB2390-100-29
Arsenic limit test apparatus
Digestion Systems Trace Element Detection Apparatus
This apparatus is used for the determination of the limit of arsenic.
Complete unit consists of three parts with attachment and tension springs.