Ramsbottom coking bulb
Supplier: DWK Life Sciences
201-1419EA 823 EUR
201-1419
Ramsbottom coking bulb
Petroleum Testing Apparatus
Ramsbottom coking bulbs are used in the determination of carbon residue in petroleum products.
Certifikace: ASTM D524. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.