Triglycerol ≥60% (by HPLC), Technical Grade, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Polyglycerin-3, 1,3-Bis(2,3-dihydroxypropyl)-2-propanol, Triglycerine

17782-250ML 17782-50ML
SIAL17782-250MLEA 363 EUR
SIAL17782-250ML SIAL17782-50ML
Triglycerol ≥60% (by HPLC), Technical Grade, Sigma-Aldrich®
Triglycerol

Triglycerol was used in the synthesis of (poly)glycerol monoethers. It was also used to enhance the chromatographic behavior of the pesticides in tea matrices.

Formula: C₉H₂₀O₇
MW: 240.25 g/mol
Density: 1.30 g/mL (20 °C) (lit.)
MDL Number: MFCD00084755
CAS Number: 20411-31-8

Order Now

Specification Test Results

assay ≥60% (HPLC)
impurities ≤20% Di- and Tetraglycerol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR