ESD safety shoes, lace-up, A 105
Supplier: ATLAS - the shoe company
74700-S3-38 74700-S3-36 74700-S3-37 74700-S3-39 74700-S3-40 74700-S3-41 74700-S3-42 74700-S3-43 74700-S3-44 74700-S3-45 74700-S3-46 74700-S3-47 74700-S3-48 74700-S3-49
114-8449EA 0 EUR
114-8449 114-8537 114-8448 114-8450 114-8451 114-8452 114-8453 114-8454 114-8455 114-8456 114-8457 114-8458 114-8459 114-8460
ESD safety shoes, lace-up, A 105
Shoes and Boots
Sneaker style ESD safety shoes made from full-grained calf leather and sneaker line sole (oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopaedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • XP® metal free puncture-resistance
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Brown
Certifikace: EN 61340 (ESD); EN ISO 20345; S3