Florisil®, BAKER ANALYZED® (60-100 Mesh), activated, J.T.Baker®
Supplier: Avantor
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
3372-07EA 312.79 EUR
3372-07 3372-08
Florisil®, BAKER ANALYZED® (60-100 Mesh), activated, J.T.Baker®
Magnesium silicate
For chromatographic cleanup of pesticide residue.
  • Activated at 675 °C
Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 Kg/L
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use ≤ 10 %
Mesh - On U.S. No. 60 Sieve ≤ 10 %
Mesh - Thru U.S. No. 100 Sieve ≤ 10 %
Product may contain black particles that are produced during the manufacturing process.