Reusable earplugs, whisper+ detec
Supplier: Uvex
uvex whisper+
UVEX2111.239EA 138 EUR
UVEX2111.239
Reusable earplugs, whisper+ detec
Earplugs Reusable Earplugs
Earplugs with cord are conveniently at hand, whenever required. Suitable for use in dirty working environments.
  • Can be worn comfortably around the neck
  • Hygienic storage in an individual carrying case
  • Dirt-repellent surface can be cleaned using soap and water
  • Maintain the wearer's comfort and safety
Suitable for low noise areas.
SNR: 27 dB
Certifikace: EN 352-2:2002