Chemical resistant gloves, uvex rubiflex S NB27S
Supplier: UVEX
9893002 9893005 9893059 9893730
UVEX9893002EA 229 EUR
UVEX9893002 UVEX9893005 UVEX9893059 UVEX9893730
Chemical resistant gloves, uvex rubiflex S NB27S
Gloves
These chemical protection gloves feature a reinforced cotton interlock lining with a full NBR coating. This makes the gloves suitable for many applications in which chemical, heat or abrasion resistance is required.
  • Good resistance to chemicals, acids, alkalis, mineral oils and solvents
  • Good heat insulation with reinforced supporting material
  • Extremely high flexibility
  • Good tactile feel
  • Outstanding wearer comfort and ergonomic fit
Typical applications include: Chemical industry, sandblasting, automotive industry, metal working and machine and tool construction.
Certifikace: EN ISO 374-1:2016 Type A (JKOPT), EN 388:2016 (2121X).
Food contact approved.