AOCS-Tintometer colour reference standards, Supelco®
Supplier: Merck
134260-500ML
134270-500ML
SIAL134260-500MLEA
278
EUR
SIAL134260-500ML
SIAL134270-500ML
AOCS-Tintometer colour reference standards, Supelco®
Standards
Colour Reference and Spectroscopy Standards
AOCS-Tintometer Colour Standard for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
- Certified under ISO 9001 Quality system.
- Full Traceability to International Standards.
- 500mL Pack Size supplied with MSDS
- 12 months shelf life.
- Tamper evident security packaging