Safety ankle boots, lace-up, Black Eagle® safety 54 low
Supplier: HAIX SCHUHE
61000810 61000810.5 61000811 61000811.5 61000812 61000812.5 61000813 61000813.5 61000814 61000814.5 61000815 6100084 6100084.5 6100085 6100085.5 6100086 6100086.5 6100087 6100087.5
HAIX61000810EA 207 EUR
HAIX61000810 HAIX61000810.5 HAIX61000811 HAIX61000811.5 HAIX61000812 HAIX61000812.5 HAIX61000813 HAIX61000813.5 HAIX61000814 HAIX61000814.5 HAIX61000815 HAIX6100084 HAIX6100084.5 HAIX6100085 HAIX6100085.5 HAIX6100086 HAIX6100086.5 HAIX6100087 HAIX6100087.5
Safety ankle boots, lace-up, Black Eagle® safety 54 low
Shoes and Boots
This shoe is an extremely lightweight safety ankle boot made from hydrophobic, breathable and waterproof leather and is flexible for alternating inside and outside use.
  • Abrasion-resistant lining with optimised climate comfort
  • Excellent thermal insulation
  • Antistatic
  • Protective metal through sole
  • Anti-slip resistance
  • Abrasion resistant TPU toe cap
  • Pulling strap and no side seams
This shoe reduces the heating effect of the upper leather as sunlight is reflected by the leather, keeping the leather and the feet cooler. Comfortable, cushioning, moisture-repellent and anti-bacterial insert. Separate heel shape for good cushioning and foot insertion.
Modern sporty sole constructed with rubber/PU street/terrain tread, abrasion-proof, to enhance toe-to-heel support, ideal for cold weather conditions. The non-marking outer sole is resistant to heat, oil and gasoline. Anatomically formed and ultralight protective toe cap made of fibre-reinforced plastic for highest safety requirements.
Certifikace: Electrostatically conductive footwear tested according to DIN EN 61340-4-3:2002.
Informace pro objednávání: A yellow insole (wide) and blue insole (narrow) can be ordered separately.