Mini centrifuge / Vortex mixer, FVL-2400N Combi-Spin
Supplier: Biosan
BSAN010202-AAAEA 468 EUR
BSAN010202-AAA
Mini centrifuge / Vortex mixer, FVL-2400N Combi-Spin
Centrifuges Microcentrifuges
Mini-centrifuge/vortex Combi–Spin FVL-2400N is specially designed for genetic engineering research (for PCR–diagnostics experiments). Units can be used in microbiological, biochemical, clinical laboratories and industrial biotechnological laboratories.
Combi–Spin provides simultaneous mixing and separation of samples, using centrifuge and mixing modules, located on the common spin–module. FVL-2400N is provided with protection mechanism that stops the rotor motion when the lid is opened.