ESD safety ankle boots, lace-up, uvex 2 xenova®, 9556
Supplier: Uvex
uvex 2 xenova®
UVEX9556.7/49EA 117 EUR
UVEX9556.7/49 UVEX9556.8/39 UVEX9556.7/47 UVEX9556.8/38 UVEX9556.7/48 UVEX9556.9/49 UVEX9556.7/45 UVEX9556.9/48 UVEX9556.7/46 UVEX9556.9/47 UVEX9556.9/46 UVEX9556.7/43 UVEX9556.7/44 UVEX9556.9/45 UVEX9556.9/44 UVEX9556.7/41 UVEX9556.9/43 UVEX9556.7/42 UVEX9556.8/52 UVEX9556.9/42 UVEX9556.9/41 UVEX9556.7/40 UVEX9556.8/51 UVEX9556.8/50 UVEX9556.9/40 UVEX9556.7/38 UVEX9556.7/39 UVEX9556.8/49 UVEX9556.9/39 UVEX9556.8/48 UVEX9556.9/38 UVEX9556.8/47 UVEX9556.8/46 UVEX9556.8/45 UVEX9556.8/44 UVEX9556.8/43 UVEX9556.7/52 UVEX9556.8/42 UVEX9556.8/41 UVEX9556.7/50 UVEX9556.9/52 UVEX9556.8/40 UVEX9556.7/51 UVEX9556.9/51 UVEX9556.9/50
ESD safety ankle boots, lace-up, uvex 2 xenova®, 9556
Shoes and Boots
Metal-free, lightweight, flexible and robust safety shoe made from water-repellent, full-grain smooth leather upper material, soft padded tongue and collar, and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Anti-static comfort insole with moisture control system; orthopaedic insoles
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/blue
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC