Methyl Indole-2-carboxylate 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Methyl 1H-indole-2-carboxylate, Indole-2-carboxylic Acid Methyl Ester
H66609.03 H66609.06
H66609.03EA 34.3 EUR
H66609.03 H66609.06
Methyl Indole-2-carboxylate 97%
Methyl Indole-2-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00460779
CAS Number: 1202-04-6
EINECS: 214-861-7
Order Now
Specification Test Results
Purity Min. 96%