J.T.Baker® BAKERBOND®, Wide-Pore Carboxy-Sulfon Ion Exchnge Media
Supplier: Avantor
J.T.Baker® BAKERBOND®
7184-02EA 0 EUR
7184-02 7252-00
J.T.Baker® BAKERBOND®, Wide-Pore Carboxy-Sulfon Ion Exchnge Media
Chromatography Sorbents HPLC Sorbents
Prep-scale packing is intended for use at low pressure with a peristaltic pump.
  • Weak-strong cation exchanger