Avantor® Hichrom, HPLC Columns, C18, 5 μm
Supplier: Avantor
HI-5C18-100AEA 801 EUR
HI-5C18-100A HI-5C18-100P HICHHI-5C18-10CP HI-5C18-150A HI-5C18-150AF HI-5C18-150AS HI-5C18-250A HI-5C18-250AF HI-5C18-250AS HI-5C18-250P HI-5C18-50P HI-5C18-50SP1 HI-5C18-75P HI-5C18-100P3 HI-5C18-100SP1 HI-5C18-10C5 HI-5C18-10CM5 HI-5C18-10CP3 HI-5C18-150P3 HI-5C18-150SP1 HI-5C18-250P3 HI-5C18-250SP1 HI-5C18-50P3 HI-5C18-300AE HI5C1810C5SK HI-5C18-125AF HI-5C18-200A HI-5C18-300A HI-5C18-300AF
Avantor® Hichrom, HPLC Columns, C18, 5 μm
Chromatography Columns
These Avantor® Hichrom C18 columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra pure silica, advanced bonding technology, superior column specification and comprehensive batch validation all contribute to the columns’ excellent reproducibility.
  • Ultra pure, base deactivated porous silica
  • Suitable for acidic, basic and neutral molecules
  • Particle sizes 3,5 and 5 μm available
Combining ultra pure silica with advanced bonding technology results in a densely bonded silica that is both highlyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Carbon load 15%
Endcapped Yes
Surface area 250 m²/g