SYBR® Green I 10000x DMSO stain_nucleic acid gel
Supplier: Biotium
Warning
Synonyms: Thiazole Green
40086-0.5ML 40086-1ML
40086-0.5MLEA 228 EUR
40086-0.5ML 40086-1ML
SYBR® Green I 10000x DMSO stain_nucleic acid gel
SYBR® Green I
Thiazole green, which is structurally identical to SYBR® green I nucleic acid gel stain, is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose, polyacrylamide gels and qPCR.
  • Color & Form Orange solution
  • Ex/Em 497/520nm (with DNA)
Thiazole green can also be used to detect ssDNA and RNA in denaturing agarose/formaldehyde and polyacrylamide/urea gels without any prewashing steps, although the sensitivity is lower. Thiazole green has the identical spectral properties as SYBR® green I (Ex/Em 497/520 nm with DNA).
SYBR is a registered trademark of Thermo Fisher Scientific.
Formula: C₃₂H₃₇N₄S
MW: 509.73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00284715
CAS Number: 163795-75-3