Polyacrylic acid sodium salt solution, M.W. ∼8000 45% in water

Supplier: Merck
Warning

Synonyms: Poly(sodium acrylate), Poly(sodium Acrylate), Acrylic Acid Sodium Salt Homopolymer, PAAS, Sodium Polyacrylate

M-Clarity™ quality level = MQ200

416029-500ML 416029-100ML
SIAL416029-500MLEA 317 EUR
SIAL416029-500ML SIAL416029-100ML
Polyacrylic acid sodium salt solution, M.W. ∼8000 45% in water
Polyacrylic acid sodium salt
Formula: [CH₂CH(CO₂Na)]n
Density: 1.3 g/cm³ (25 °C)
MDL Number: MFCD00147949
CAS Number: 9003-04-7

Order Now

Specification Test Results

Appearance (Colour) Colourless
Appearance (Form) Viscous Liquid
Proton NMR Spectrum Conforms to Structure
Residue on Evaporation 43.0 - 47.0% wt

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR