Tube Carriers
Supplier: Hamilton
HAMI173410EA 381 EUR
HAMI173410
Tube Carriers
Carriers Transport Carriers
These tube carriers carries 24 and 32 tubes and occupies one track.
For tubes with 11 - 14 mm outer diameter, 60 - 120 mm high.