Phloxin B 3% in aqueous solution for histology, for cytology
Supplier: RAL (REACTIFS RAL SA)
Synonyms: CI 45410
RALA350750-1000EA 47.82 EUR
RALA350750-1000
Phloxin B 3% in aqueous solution for histology, for cytology
Phloxin B
Formula: C₂₀H₂Br₄Cl₄Na₂O₅
MW: 829,6344 g/mol
MDL Number: MFCD00070627
EINECS: 242-355-6
IVDLogo