Hematoxylin stabilised
Supplier: RAL (REACTIFS RAL SA)
720-0360EA 103.3 EUR
720-0360
Hematoxylin stabilised
Hematoxylin
Formula: C₁₆H₁₄O₆
MW: 302,28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo