Hemaperls kit for haematology
Supplier: RAL (REACTIFS RAL SA)
720-1184EA
106
EUR
720-1184
Hemaperls kit for haematology
Hemaperls kit for haematology
Perls Staining for the Detection of Haemosiderin (Ferric salts - Prussian Blue)
In an acid environment, ferric ions react with potassium ferrocyanide what leads to a precipitate called Ferric Ferrocyanide (Prussian blue). It points out the presence of a pathological pigment: Haemosiderin.