Diethylenetriaminepentaacetic acid, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
86179.180EA
105
EUR
86179.180
Diethylenetriaminepentaacetic acid, AnalaR NORMAPUR® analytical reagent
Diethylenetriaminepentaacetic acid
Specification Test Results
Appearance | White crystalline powder |
Assay (complexometric on dried basis) | Min 99,0 % |
Heavy metals (as Pb) | Max 0,0010% |
Fe (Iron) | Max 0,0010% |
Water | Max 0,50% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...