IGEPAL® CA-630, viscous liquid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Octylphenyl-polyethylene glycol
M-Clarity™ quality level = MQ200
IGEPAL® CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes. It is chemically indistinguishable from Nonidet P-40.[
IGEPAL® CA-630 has been used:
• As a component of grinding buffer along with lithium dodecyl sulphate, tris-HCl, EDTA, NaCl, sodium deoxycholate, dithiothreitol and thiourea in RNA extraction process
• Along with Tris- HCl finds its application in incubation of Tachyzoites cultured in vero cells
• As a component of nuclear extract buffer D+ containing HEPES, glycerol, KCl and EDTA for gel shift analysis
• As a component of PI buffer along with NaH2PO4 to wash slides in FISH (fluorescence in situ hybridization) procedure
Formula:
(C₂H₄O)nC₁₄H₂₂O Flash Pt: 251 °C |
MDL Number:
MFCD00132505 CAS Number: 9002-93-1 UN: 3082 ADR: 9,III |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...