Magnesium chloride 1 mol/l 2 N

Supplier: VWR Chemicals

85966.230EA 93.1 EUR
85966.230
Magnesium chloride 1 mol/l 2 N
Magnesium chloride
Formula: MgCl₂
MW: 95,21 g/mol
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Specification Test Results

Titer 0,999 --> 1,001 Mol/l
Titer (HCl) 0.0110 --> 0.0122 Mol/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR