ESD safety sandals, Velcro® fastening, uvex 2 trend, 6946
Supplier: Uvex
uvex 2 trend
UVEX6946.3/52EA 76.9 EUR
UVEX6946.3/52 UVEX6946.1/51 UVEX6946.2/41 UVEX6946.3/50 UVEX6946.2/40 UVEX6946.1/50 UVEX6946.3/51 UVEX6946.1/39 UVEX6946.1/38 UVEX6946.2/47 UVEX6946.3/38 UVEX6946.3/39 UVEX6946.2/46 UVEX6946.2/49 UVEX6946.2/48 UVEX6946.2/43 UVEX6946.1/52 UVEX6946.2/42 UVEX6946.2/45 UVEX6946.2/44 UVEX6946.3/41 UVEX6946.2/50 UVEX6946.3/42 UVEX6946.2/52 UVEX6946.1/40 UVEX6946.3/40 UVEX6946.2/51 UVEX6946.2/39 UVEX6946.1/49 UVEX6946.1/46 UVEX6946.3/49 UVEX6946.1/45 UVEX6946.1/48 UVEX6946.3/47 UVEX6946.2/38 UVEX6946.1/47 UVEX6946.3/48 UVEX6946.1/42 UVEX6946.3/45 UVEX6946.1/41 UVEX6946.3/46 UVEX6946.3/43 UVEX6946.1/44 UVEX6946.1/43 UVEX6946.3/44
ESD safety sandals, Velcro® fastening, uvex 2 trend, 6946
Shoes and Boots
ESD sporty S1P safety sandals with wide fit, made from microvelour upper and a dual-density PU sole.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable microvelour upper
  • Penetration resistant steel midsole and steel toe cap
  • Double, individually adjustable velcro fastening
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • High levels of wearer comfort
Safety sandals feature an ergonomically designed outsole that offers excellent slip resistance, as well as a stabilising uvex anti-twist heel cap designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances. Removable, anti-static, comfortable insole, with a moisture transport system and additional shock absorption around the heel and forefoot.
Colour: Black/grey
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC